The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003566.3 | NP_003557.2 | 16 - 17 | 2313 | 76 |
XM_011538814.2 | XP_011537116.1 | 17 - 18 | 2419 | 76 | |
XM_011538815.2 | XP_011537117.1 | 16 - 17 | 2130 | 76 | |
XM_017020018.1 | XP_016875507.1 | 16 - 17 | 2081 | 76 | |
GenBank mRNA | AK292008.1 | - | 16 - 17 | 2232 | 76 |
AK307918.1 | - | 16 - 17 | 2234 | 76 | |
X78998.1 | - | 16 - 17 | 2119 | 76 |