The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303058.1 | NP_001289987.1 | 1 - 2 | 249 | 66 |
NM_001303059.1 | NP_001289988.1 | 1 - 2 | 249 | 66 | |
NM_182547.3 | NP_872353.2 | 1 - 2 | 249 | 66 | |
GenBank mRNA | AK074557.1 | - | 1 - 2 | 185 | 66 |
AK290125.1 | - | 1 - 2 | 195 | 66 | |
AY191223.1 | - | 1 - 2 | 168 | 66 | |
BC035467.1 | - | 1 - 2 | 172 | 66 | |
BC052641.1 | - | 1 - 2 | 182 | 66 | |
BC053583.1 | - | 1 - 2 | 182 | 66 | |
BC057851.1 | - | 1 - 2 | 185 | 66 | |
BX647965.1 | - | 1 - 2 | 165 | 66 | |
DA419871.1 | - | 1 - 2 | 249 | 66 |