Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALKBH3
Entrez Gene ID
221120
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
Gene Aliases
ABH3, DEPC-1, DEPC1, PCA1, hABH3
Chromosome Location
Chr.11: 43880807 - 43920275 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139178.3 NP_631917.1 9 - 10 1211 76
GenBank mRNA AB042029.1 - 9 - 10 1174 76
AK315266.1 - 9 - 10 1278 76
BC015155.1 - 4 - 5 497 76
BC067257.1 - 9 - 10 1208 76
BC103812.1 - 9 - 10 949 76
BC103813.1 - 9 - 10 949 76
BC103814.2 - 9 - 10 949 76
CD364334.1 - 1 - 2 76

View Full Product Details