The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000358.2 | NP_000349.1 | 10 - 11 | 1560 | 73 |
GenBank mRNA | AK093916.1 | - | 10 - 11 | 1415 | 73 |
AK094055.1 | - | 11 - 12 | 1491 | 73 | |
AK094581.1 | - | 10 - 11 | 1504 | 73 | |
AK222833.1 | - | 10 - 11 | 1468 | 73 | |
AK298603.1 | - | 5 - 6 | 780 | 73 | |
AK303643.1 | - | 10 - 11 | 1792 | 73 | |
BC000097.2 | - | 10 - 11 | 1453 | 73 | |
BC004972.1 | - | 10 - 11 | 1448 | 73 | |
BT009820.1 | - | 10 - 11 | 1399 | 73 | |
KU178357.1 | - | 10 - 11 | 1399 | 73 | |
KU178358.1 | - | 8 - 9 | 913 | 73 | |
M77349.1 | - | 10 - 11 | 1446 | 73 |