The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000358.2 | NP_000349.1 | 8 - 9 | 1286 | 103 |
GenBank mRNA | AB209598.1 | - | 2 - 3 | 4048 | 103 |
AK093916.1 | - | 8 - 9 | 1141 | 103 | |
AK094055.1 | - | 9 - 10 | 1217 | 103 | |
AK094581.1 | - | 8 - 9 | 1230 | 103 | |
AK222833.1 | - | 8 - 9 | 1194 | 103 | |
AK298603.1 | - | 3 - 4 | 506 | 103 | |
AK303643.1 | - | 8 - 9 | 1518 | 103 | |
BC000097.2 | - | 8 - 9 | 1179 | 103 | |
BC004972.1 | - | 8 - 9 | 1174 | 103 | |
BT009820.1 | - | 8 - 9 | 1125 | 103 | |
KU178357.1 | - | 8 - 9 | 1125 | 103 | |
M77349.1 | - | 8 - 9 | 1172 | 103 |