Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS54
Entrez Gene ID
51542
Gene Name
VPS54, GARP complex subunit
Gene Aliases
HCC8, PPP1R164, SLP-8p, VPS54L, WR, hVps54L
Chromosome Location
Chr.2: 63892149 - 64019421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005739.1 NP_001005739.1 13 - 14 1992 132
NM_016516.2 NP_057600.2 13 - 14 2028 132
XM_006712029.3 XP_006712092.1 12 - 13 1888 132
XM_017004316.1 XP_016859805.1 12 - 13 1708 132
GenBank mRNA AK002205.1 - 1 - 2 133 132
AK025545.1 - 7 - 8 920 132
AK124436.1 - 2 - 3 3176 132
AK292669.1 - 14 - 15 2036 132
AK292889.1 - 14 - 15 2087 132
AY444798.1 - 12 - 13 1885 132
BC030275.1 - 13 - 14 1995 132
BC041868.1 - 10 - 11 1996 132

View Full Product Details