Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX16
Entrez Gene ID
9409
Gene Name
peroxisomal biogenesis factor 16
Gene Aliases
PBD8A, PBD8B
Chromosome Location
Chr.11: 45909669 - 45918123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004813.2 NP_004804.1 10 - 11 1269 113
XM_011520474.1 XP_011518776.1 10 - 11 1008 113
GenBank mRNA AB016531.1 - 10 - 11 994 113
AF118240.1 - 10 - 11 1201 113
BC004356.1 - 10 - 11 967 113
BM746616.1 - 2 - 3 89 113

View Full Product Details