Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX16
Entrez Gene ID
9409
Gene Name
peroxisomal biogenesis factor 16
Gene Aliases
PBD8A, PBD8B
Chromosome Location
Chr.11: 45909669 - 45918123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004813.2 NP_004804.1 1 - 2 427 75
NM_057174.2 NP_476515.1 1 - 2 427 75
GenBank mRNA AB016531.1 - 1 - 2 152 75
AF118240.1 - 1 - 2 359 75
BC000467.2 - 1 - 2 133 75
BC004356.1 - 1 - 2 125 75
CN285513.1 - 1 - 2 434 75

View Full Product Details