Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX16
Entrez Gene ID
9409
Gene Name
peroxisomal biogenesis factor 16
Gene Aliases
PBD8A, PBD8B
Chromosome Location
Chr.11: 45909669 - 45918123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004813.2 NP_004804.1 2 - 3 455 69
NM_057174.2 NP_476515.1 2 - 3 455 69
GenBank mRNA AB016531.1 - 2 - 3 180 69
AF118240.1 - 2 - 3 387 69
BC000467.2 - 2 - 3 161 69
BC004356.1 - 2 - 3 153 69
CN285513.1 - 2 - 3 462 69

View Full Product Details