The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004813.2 | NP_004804.1 | 3 - 4 | 542 | 82 |
NM_057174.2 | NP_476515.1 | 3 - 4 | 542 | 82 | |
XM_011520474.1 | XP_011518776.1 | 3 - 4 | 281 | 82 | |
GenBank mRNA | AB016531.1 | - | 3 - 4 | 267 | 82 |
AF118240.1 | - | 3 - 4 | 474 | 82 | |
BC000467.2 | - | 3 - 4 | 248 | 82 | |
BC004356.1 | - | 3 - 4 | 240 | 82 | |
CN285513.1 | - | 3 - 4 | 549 | 82 |