Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS54
Entrez Gene ID
51542
Gene Name
VPS54, GARP complex subunit
Gene Aliases
HCC8, PPP1R164, SLP-8p, VPS54L, WR, hVps54L
Chromosome Location
Chr.2: 63892149 - 64019421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005739.1 NP_001005739.1 1 - 2 148 98
NM_016516.2 NP_057600.2 1 - 2 148 98
XM_006712029.3 XP_006712092.1 1 - 2 116 98
XM_017004316.1 XP_016859805.1 1 - 2 70 98
GenBank mRNA AK292889.1 - 2 - 3 243 98
AL514510.3 - 1 - 2 254 98
BC030275.1 - 1 - 2 151 98

View Full Product Details