The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001695.4 | NP_001686.1 | 8 - 9 | 884 | 72 |
GenBank mRNA | AK000064.1 | - | 8 - 9 | 812 | 72 |
AK298617.1 | - | 6 - 7 | 559 | 72 | |
AK308632.1 | - | 7 - 8 | 645 | 72 | |
BC010960.1 | - | 8 - 9 | 810 | 72 | |
BX648079.1 | - | 8 - 9 | 801 | 72 | |
J05682.1 | - | 7 - 8 | 534 | 72 | |
X69151.1 | - | 8 - 9 | 805 | 72 |