The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270453.1 | NP_001257382.1 | 10 - 11 | 1260 | 86 |
NM_001270454.1 | NP_001257383.1 | 13 - 14 | 1570 | 86 | |
NM_007014.4 | NP_008945.2 | 14 - 15 | 1623 | 86 | |
NM_199424.2 | NP_955456.1 | 3 - 4 | 715 | 86 | |
XM_011522823.2 | XP_011521125.1 | 15 - 16 | 1653 | 86 | |
XM_011522825.1 | XP_011521127.1 | 13 - 14 | 1534 | 86 | |
XM_011522826.2 | XP_011521128.1 | 10 - 11 | 1424 | 86 | |
XM_017022879.1 | XP_016878368.1 | 15 - 16 | 1727 | 86 | |
XM_017022880.1 | XP_016878369.1 | 15 - 16 | 1659 | 86 | |
XM_017022881.1 | XP_016878370.1 | 15 - 16 | 1703 | 86 | |
GenBank mRNA | AK126332.1 | - | 3 - 4 | 700 | 86 |
AK295070.1 | - | 6 - 7 | 615 | 86 | |
AK295701.1 | - | 11 - 12 | 1319 | 86 | |
AK300266.1 | - | 10 - 11 | 1260 | 86 | |
AK312792.1 | - | 13 - 14 | 1550 | 86 | |
AK316097.1 | - | 4 - 5 | 569 | 86 | |
BC013645.1 | - | 12 - 13 | 1457 | 86 | |
BC064531.1 | - | 13 - 14 | 1509 | 86 | |
BU167468.1 | - | - | 116 | 86 | |
JN712744.1 | - | 13 - 14 | 1540 | 86 | |
U96114.1 | - | 14 - 15 | 1555 | 86 |