The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270453.1 | NP_001257382.1 | 15 - 16 | 1786 | 73 |
NM_001270454.1 | NP_001257383.1 | 18 - 19 | 2096 | 73 | |
NM_007014.4 | NP_008945.2 | 19 - 20 | 2149 | 73 | |
NM_199424.2 | NP_955456.1 | 8 - 9 | 1241 | 73 | |
XM_011522823.2 | XP_011521125.1 | 20 - 21 | 2179 | 73 | |
XM_011522825.1 | XP_011521127.1 | 18 - 19 | 2060 | 73 | |
XM_011522826.2 | XP_011521128.1 | 15 - 16 | 1950 | 73 | |
XM_017022879.1 | XP_016878368.1 | 20 - 21 | 2253 | 73 | |
XM_017022880.1 | XP_016878369.1 | 20 - 21 | 2185 | 73 | |
XM_017022881.1 | XP_016878370.1 | 20 - 21 | 2229 | 73 | |
GenBank mRNA | AK126332.1 | - | 8 - 9 | 1226 | 73 |
AK295070.1 | - | 11 - 12 | 1141 | 73 | |
AK295701.1 | - | 16 - 17 | 1845 | 73 | |
AK300266.1 | - | 15 - 16 | 1786 | 73 | |
AK312792.1 | - | 18 - 19 | 2076 | 73 | |
AK316097.1 | - | 9 - 10 | 1095 | 73 | |
BC013645.1 | - | 17 - 18 | 1983 | 73 | |
BC064531.1 | - | 18 - 19 | 2035 | 73 | |
JN712744.1 | - | 18 - 19 | 2066 | 73 | |
U96114.1 | - | 19 - 20 | 2081 | 73 |