The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270453.1 | NP_001257382.1 | 5 - 6 | 731 | 126 |
NM_001270454.1 | NP_001257383.1 | 8 - 9 | 1041 | 126 | |
NM_001270455.1 | NP_001257384.1 | 8 - 9 | 1041 | 126 | |
NM_007014.4 | NP_008945.2 | 9 - 10 | 1094 | 126 | |
XM_011522823.2 | XP_011521125.1 | 10 - 11 | 1124 | 126 | |
XM_011522825.1 | XP_011521127.1 | 8 - 9 | 1005 | 126 | |
XM_011522826.2 | XP_011521128.1 | 5 - 6 | 895 | 126 | |
XM_017022879.1 | XP_016878368.1 | 10 - 11 | 1198 | 126 | |
XM_017022880.1 | XP_016878369.1 | 10 - 11 | 1130 | 126 | |
XM_017022881.1 | XP_016878370.1 | 10 - 11 | 1174 | 126 | |
GenBank mRNA | AK295701.1 | - | 6 - 7 | 790 | 126 |
AK300266.1 | - | 5 - 6 | 731 | 126 | |
AK312792.1 | - | 8 - 9 | 1021 | 126 | |
BC000108.2 | - | 7 - 8 | 943 | 126 | |
BC013645.1 | - | 7 - 8 | 928 | 126 | |
BC064531.1 | - | 8 - 9 | 980 | 126 | |
BX372347.2 | - | 1 - 2 | 126 | ||
JN712744.1 | - | 8 - 9 | 1011 | 126 | |
U96114.1 | - | 9 - 10 | 1026 | 126 |