Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REEP2
Entrez Gene ID
51308
Gene Name
receptor accessory protein 2
Gene Aliases
C5orf19, SGC32445, SPG72, Yip2d
Chromosome Location
Chr.5: 138439001 - 138446969 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016606.3 NP_057690.2 5 - 6 629 57
NR_073448.1 5 - 6 704 57
GenBank mRNA AK293480.1 - 5 - 6 603 57
AY562240.1 - 5 - 6 421 57
BC006218.2 - 5 - 6 543 57

View Full Product Details