Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REEP2
Entrez Gene ID
51308
Gene Name
receptor accessory protein 2
Gene Aliases
C5orf19, SGC32445, SPG72, Yip2d
Chromosome Location
Chr.5: 138439001 - 138446969 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271803.1 NP_001258732.1 7 - 8 905 67
NM_016606.3 NP_057690.2 7 - 8 899 67
NR_073448.1 7 - 8 974 67
NR_073449.1 7 - 8 980 67
GenBank mRNA AF070551.1 - 3 - 4 343 67
AF251041.1 - 6 - 7 679 67
AK056193.1 - 7 - 8 861 67
AK223611.1 - 7 - 8 846 67
AK290547.1 - 7 - 8 795 67
AK293480.1 - 7 - 8 873 67
AY562240.1 - 7 - 8 691 67
BC006218.2 - 7 - 8 813 67

View Full Product Details