The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015702.2 | NP_056517.1 | 7 - 8 | 898 | 88 |
GenBank mRNA | AF131802.1 | - | 7 - 8 | 788 | 88 |
AF161510.1 | - | 7 - 8 | 850 | 88 | |
AK313284.1 | - | 7 - 8 | 797 | 88 | |
BC000932.2 | - | 7 - 8 | 847 | 88 | |
BC010894.1 | - | 7 - 8 | 821 | 88 | |
BC022859.1 | - | 7 - 8 | 844 | 88 | |
BC023995.1 | - | 7 - 8 | 812 | 88 |