Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSG3
Entrez Gene ID
1830
Gene Name
desmoglein 3
Gene Aliases
CDHF6, PVA
Chromosome Location
Chr.18: 31447769 - 31478702 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001944.2 NP_001935.2 9 - 10 1385 157
XM_011525850.1 XP_011524152.1 9 - 10 1385 157
GenBank mRNA AK290367.1 - 9 - 10 1385 157
M76482.1 - 9 - 10 1359 157

View Full Product Details