Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR14
Entrez Gene ID
64419
Gene Name
myotubularin related protein 14
Gene Aliases
C3orf29
Chromosome Location
Chr.3: 9649433 - 9702394 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077525.2 NP_001070993.1 3 - 4 567 67
NM_001077526.2 NP_001070994.1 3 - 4 567 67
NM_022485.4 NP_071930.2 3 - 4 567 67
XM_005265400.1 XP_005265457.1 3 - 4 540 67
XM_011534012.1 XP_011532314.1 2 - 3 336 67
XM_011534013.1 XP_011532315.1 2 - 3 292 67
XM_017007034.1 XP_016862523.1 4 - 5 737 67
XM_017007035.1 XP_016862524.1 4 - 5 737 67
XM_017007036.1 XP_016862525.1 4 - 5 736 67
XM_017007037.1 XP_016862526.1 4 - 5 737 67
XM_017007038.1 XP_016862527.1 3 - 4 540 67
XM_017007039.1 XP_016862528.1 4 - 5 736 67
XM_017007040.1 XP_016862529.1 4 - 5 737 67
XM_017007041.1 XP_016862530.1 2 - 3 336 67
XM_017007042.1 XP_016862531.1 3 - 4 540 67
XM_017007043.1 XP_016862532.1 2 - 3 336 67
GenBank mRNA AF289603.1 - 3 - 4 530 67
AK026058.1 - 2 - 3 134 67
AK074792.1 - 3 - 4 538 67
AL713695.1 - 3 - 4 488 67
BC001674.1 - 3 - 4 472 67
BC025952.1 - 3 - 4 517 67
BC035690.1 - 3 - 4 479 67
BC050626.1 - 3 - 4 490 67
DQ630520.1 - 3 - 4 416 67

View Full Product Details