Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MTMR14
- Entrez Gene ID
-
64419
- Gene Name
- myotubularin related protein 14
- Gene Aliases
-
C3orf29
- Chromosome Location
-
Chr.3: 9649433 - 9702394 on Build GRCh38
View Full Product Details