The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077525.2 | NP_001070993.1 | 2 - 3 | 456 | 75 |
NM_001077526.2 | NP_001070994.1 | 2 - 3 | 456 | 75 | |
NM_022485.4 | NP_071930.2 | 2 - 3 | 456 | 75 | |
XM_005265400.1 | XP_005265457.1 | 2 - 3 | 429 | 75 | |
XM_017007034.1 | XP_016862523.1 | 3 - 4 | 626 | 75 | |
XM_017007035.1 | XP_016862524.1 | 3 - 4 | 626 | 75 | |
XM_017007036.1 | XP_016862525.1 | 3 - 4 | 625 | 75 | |
XM_017007037.1 | XP_016862526.1 | 3 - 4 | 626 | 75 | |
XM_017007038.1 | XP_016862527.1 | 2 - 3 | 429 | 75 | |
XM_017007039.1 | XP_016862528.1 | 3 - 4 | 625 | 75 | |
XM_017007040.1 | XP_016862529.1 | 3 - 4 | 626 | 75 | |
XM_017007042.1 | XP_016862531.1 | 2 - 3 | 429 | 75 | |
XM_017007044.1 | XP_016862533.1 | 2 - 3 | 423 | 75 | |
XM_017007045.1 | XP_016862534.1 | 2 - 3 | 398 | 75 | |
GenBank mRNA | AF289603.1 | - | 2 - 3 | 419 | 75 |
AK074792.1 | - | 2 - 3 | 427 | 75 | |
AK295891.1 | - | 2 - 3 | 384 | 75 | |
AL713695.1 | - | 2 - 3 | 377 | 75 | |
BC001674.1 | - | 2 - 3 | 361 | 75 | |
BC025952.1 | - | 2 - 3 | 406 | 75 | |
BC035690.1 | - | 2 - 3 | 368 | 75 | |
BC050626.1 | - | 2 - 3 | 379 | 75 | |
DA604126.1 | - | 2 - 3 | 429 | 75 | |
DC316206.1 | - | 2 - 3 | 458 | 75 | |
DQ630520.1 | - | 2 - 3 | 305 | 75 |