The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015032.3 | NP_055847.1 | 25 - 26 | 3125 | 99 |
XM_005266298.4 | XP_005266355.1 | 25 - 26 | 3120 | 99 | |
XM_011534999.2 | XP_011533301.1 | 25 - 26 | 3120 | 99 | |
XM_011535000.2 | XP_011533302.1 | 25 - 26 | 3120 | 99 | |
XM_011535002.2 | XP_011533304.1 | 18 - 19 | 2222 | 99 | |
XM_017020448.1 | XP_016875937.1 | 25 - 26 | 3267 | 99 | |
XM_017020449.1 | XP_016875938.1 | 25 - 26 | 3035 | 99 | |
XM_017020450.1 | XP_016875939.1 | 25 - 26 | 3035 | 99 | |
XM_017020451.1 | XP_016875940.1 | 25 - 26 | 3035 | 99 | |
XM_017020452.1 | XP_016875941.1 | 25 - 26 | 3091 | 99 | |
XM_017020453.1 | XP_016875942.1 | 25 - 26 | 3120 | 99 | |
GenBank mRNA | AB023196.2 | - | 25 - 26 | 3047 | 99 |
AK128502.1 | - | 7 - 8 | 2907 | 99 | |
AK299340.1 | - | 25 - 26 | 3001 | 99 | |
AL137201.1 | - | 25 - 26 | 3065 | 99 | |
U95825.2 | - | 25 - 26 | 3004 | 99 |