Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEATR9
Entrez Gene ID
256957
Gene Name
HEAT repeat containing 9
Gene Aliases
C17orf66
Chromosome Location
Chr.17: 35854946 - 35868891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152781.3 NP_689994.2 4 - 5 598 106
NR_135630.1 4 - 5 598 106
XM_005278233.2 XP_005278290.1 5 - 6 557 106
XM_011524586.2 XP_011522888.1 4 - 5 541 106
XM_017024424.1 XP_016879913.1 4 - 5 578 106
GenBank mRNA AK057392.1 - 4 - 5 598 106
AK301884.1 - 5 - 6 562 106
BC033734.1 - 4 - 5 546 106
BC132681.1 - 4 - 5 534 106
BC144011.1 - 4 - 5 534 106

View Full Product Details