The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005892.3 | NP_005883.2 | 18 - 19 | 2650 | 73 |
XM_006722062.3 | XP_006722125.1 | 19 - 20 | 2865 | 73 | |
XM_006722063.3 | XP_006722126.1 | 18 - 19 | 2845 | 73 | |
XM_006722064.3 | XP_006722127.1 | 19 - 20 | 2835 | 73 | |
XM_006722065.3 | XP_006722128.1 | 18 - 19 | 2815 | 73 | |
XM_006722066.3 | XP_006722129.1 | 19 - 20 | 2861 | 73 | |
XM_006722069.2 | XP_006722132.1 | 19 - 20 | 2864 | 73 | |
XM_006722070.3 | XP_006722133.1 | 18 - 19 | 2841 | 73 | |
XM_011525179.2 | XP_011523481.1 | 19 - 20 | 2865 | 73 | |
XM_011525180.2 | XP_011523482.1 | 19 - 20 | 2557 | 73 | |
XM_011525182.2 | XP_011523484.1 | 19 - 20 | 2862 | 73 | |
GenBank mRNA | AF432213.1 | - | 15 - 16 | 2124 | 73 |
AJ008112.1 | - | 3 - 4 | 566 | 73 | |
AY278319.1 | - | 18 - 19 | 2650 | 73 | |
BC001710.1 | - | 3 - 4 | 554 | 73 | |
BC021906.1 | - | 6 - 7 | 1158 | 73 | |
BC073988.1 | - | 6 - 7 | 1198 | 73 | |
BC111564.1 | - | 11 - 12 | 1463 | 73 | |
CR456759.1 | - | 3 - 4 | 527 | 73 | |
FJ534522.1 | - | 2 - 3 | 449 | 73 |