The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005892.3 | NP_005883.2 | 12 - 13 | 1432 | 84 |
XM_006722062.3 | XP_006722125.1 | 13 - 14 | 1647 | 84 | |
XM_006722063.3 | XP_006722126.1 | 12 - 13 | 1627 | 84 | |
XM_006722064.3 | XP_006722127.1 | 13 - 14 | 1617 | 84 | |
XM_006722065.3 | XP_006722128.1 | 12 - 13 | 1597 | 84 | |
XM_006722066.3 | XP_006722129.1 | 13 - 14 | 1643 | 84 | |
XM_006722069.2 | XP_006722132.1 | 13 - 14 | 1646 | 84 | |
XM_006722070.3 | XP_006722133.1 | 12 - 13 | 1623 | 84 | |
XM_011525179.2 | XP_011523481.1 | 13 - 14 | 1647 | 84 | |
XM_011525180.2 | XP_011523482.1 | 13 - 14 | 1339 | 84 | |
XM_011525182.2 | XP_011523484.1 | 13 - 14 | 1644 | 84 | |
GenBank mRNA | AF432213.1 | - | 9 - 10 | 906 | 84 |
AY278319.1 | - | 12 - 13 | 1432 | 84 | |
BC111564.1 | - | 4 - 5 | 434 | 84 |