The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002207.2 | NP_002198.2 | 24 - 25 | 2715 | 112 |
GenBank mRNA | D25303.1 | - | 25 - 26 | 2713 | 112 |
L24158.1 | - | 24 - 25 | 2557 | 112 |