Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGA9
Entrez Gene ID
3680
Gene Name
integrin subunit alpha 9
Gene Aliases
ALPHA-RLC, ITGA4L, RLC
Chromosome Location
Chr.3: 37452322 - 37819790 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002207.2 NP_002198.2 2 - 3 361 61
GenBank mRNA BC030198.1 - 2 - 3 568 61
D25303.1 - 2 - 3 359 61
L24158.1 - 2 - 3 203 61

View Full Product Details