The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001314042.1 | NP_001300971.1 | 25 - 26 | 5297 | 78 |
NM_006618.4 | NP_006609.3 | 24 - 25 | 5189 | 78 | |
XM_011509088.2 | XP_011507390.1 | 23 - 24 | 4106 | 78 | |
XM_011509090.2 | XP_011507392.1 | 25 - 26 | 4278 | 78 | |
XM_011509091.2 | XP_011507393.1 | 25 - 26 | 4015 | 78 | |
XM_011509092.2 | XP_011507394.1 | 24 - 25 | 4132 | 78 | |
GenBank mRNA | AB015348.1 | - | 4 - 5 | 784 | 78 |
AJ132440.1 | - | 24 - 25 | 4113 | 78 | |
AJ243706.1 | - | 24 - 25 | 5192 | 78 | |
AK001400.1 | - | 12 - 13 | 2238 | 78 | |
AK022553.1 | - | 15 - 16 | 2839 | 78 | |
AK074250.1 | - | 6 - 7 | 1419 | 78 | |
AK122752.1 | - | 24 - 25 | 4143 | 78 | |
AL133040.1 | - | 20 - 21 | 3442 | 78 | |
AL133048.1 | - | 13 - 14 | 2478 | 78 | |
AL133072.1 | - | 11 - 12 | 2141 | 78 | |
AL137622.1 | - | 6 - 7 | 1272 | 78 |