The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002349.3 | NP_002340.2 | 10 - 11 | 1684 | 125 |
GenBank mRNA | AB208915.1 | - | 4 - 5 | 464 | 125 |
AF011333.1 | - | 10 - 11 | 1663 | 125 | |
AF064827.1 | - | 10 - 11 | 1610 | 125 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198759.1 | NP_001185688.1 | 10 - 11 | 1684 | 125 |
NM_001198760.1 | NP_001185689.1 | 10 - 11 | 1684 | 125 | |
GenBank mRNA | AY184222.1 | - | 10 - 11 | 1610 | 125 |
AY314006.1 | - | 10 - 11 | 1610 | 125 |