The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002349.3 | NP_002340.2 | 14 - 15 | 2280 | 85 |
GenBank mRNA | AB208915.1 | - | 8 - 9 | 1060 | 85 |
AF011333.1 | - | 14 - 15 | 2259 | 85 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198759.1 | NP_001185688.1 | 14 - 15 | 2280 | 85 |
NM_001198760.1 | NP_001185689.1 | 14 - 15 | 2280 | 85 | |
GenBank mRNA | AY184222.1 | - | 14 - 15 | 2206 | 85 |
AY314006.1 | - | 14 - 15 | 2206 | 85 |