The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002349.3 | NP_002340.2 | 23 - 24 | 3214 | 136 |
GenBank mRNA | AB208915.1 | - | 17 - 18 | 1994 | 136 |
AF011333.1 | - | 23 - 24 | 3193 | 136 | |
AF064827.1 | - | 23 - 24 | 3140 | 136 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198759.1 | NP_001185688.1 | 23 - 24 | 3214 | 136 |
NM_001198760.1 | NP_001185689.1 | 23 - 24 | 3214 | 136 | |
GenBank mRNA | AY184222.1 | - | 23 - 24 | 3140 | 136 |
AY314006.1 | - | 23 - 24 | 3140 | 136 |