The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172.3 | NP_001163.1 | 5 - 6 | 732 | 141 |
GenBank mRNA | AK312484.1 | - | 5 - 6 | 648 | 141 |
AY074489.1 | - | 5 - 6 | 677 | 141 | |
BC001350.1 | - | 5 - 6 | 633 | 141 | |
BC008464.1 | - | 5 - 6 | 674 | 141 | |
BC029050.1 | - | 5 - 6 | 732 | 141 | |
CR536550.1 | - | 5 - 6 | 616 | 141 | |
D86724.1 | - | 5 - 6 | 636 | 141 | |
U75667.1 | - | 5 - 6 | 654 | 141 | |
U82256.1 | - | 5 - 6 | 632 | 141 |