The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172.3 | NP_001163.1 | 6 - 7 | 837 | 93 |
GenBank mRNA | AK312484.1 | - | 6 - 7 | 753 | 93 |
AY074489.1 | - | 6 - 7 | 782 | 93 | |
BC001350.1 | - | 6 - 7 | 738 | 93 | |
BC008464.1 | - | 6 - 7 | 779 | 93 | |
BC029050.1 | - | 6 - 7 | 837 | 93 | |
CR536550.1 | - | 6 - 7 | 721 | 93 | |
D86724.1 | - | 6 - 7 | 741 | 93 | |
U75667.1 | - | 6 - 7 | 759 | 93 | |
U82256.1 | - | 6 - 7 | 737 | 93 |