Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- OASL
- Entrez Gene ID
-
8638
- Gene Name
- 2'-5'-oligoadenylate synthetase like
- Gene Aliases
-
OASLd, TRIP-14, TRIP14, p59 OASL, p59-OASL, p59OASL
- Chromosome Location
-
Chr.12: 121018882 - 121039242 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC064330.1 |
- |
|
1354 |
|
Chr. - - [-] on Build |
65 |
View Full Product Details