Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OASL
Entrez Gene ID
8638
Gene Name
2'-5'-oligoadenylate synthetase like
Gene Aliases
OASLd, TRIP-14, TRIP14, p59 OASL, p59-OASL, p59OASL
Chromosome Location
Chr.12: 121018882 - 121039242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003733.3 NP_003724.1 5 - 6 1324 107
NM_198213.2 NP_937856.1 4 - 5 1082 107
GenBank mRNA AJ225089.1 - 5 - 6 1059 107
AK023956.1 - 5 - 6 1060 107
AK314419.1 - 5 - 6 1059 107
BC117408.1 - 5 - 6 1077 107
BC117410.1 - 5 - 6 1077 107
BM909951.1 - 3 - 4 527 107

View Full Product Details