The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261464.1 | NP_001248393.1 | 4 - 5 | 489 | 71 |
NM_001261465.1 | NP_001248394.1 | 4 - 5 | 459 | 71 | |
NM_001261466.1 | NP_001248395.1 | 4 - 5 | 411 | 71 | |
NM_003815.4 | NP_003806.3 | 4 - 5 | 459 | 71 | |
NM_207191.2 | NP_997074.1 | 4 - 5 | 459 | 71 | |
NM_207194.2 | NP_997077.1 | 4 - 5 | 459 | 71 | |
NM_207195.2 | NP_997078.1 | 4 - 5 | 459 | 71 | |
NM_207196.2 | NP_997079.1 | 4 - 5 | 459 | 71 | |
NM_207197.2 | NP_997080.1 | 4 - 5 | 459 | 71 | |
NR_048577.1 | 4 - 5 | 459 | 71 | ||
NR_048578.1 | 4 - 5 | 459 | 71 | ||
NR_048579.1 | 4 - 5 | 459 | 71 | ||
GenBank mRNA | AB209157.1 | - | 3 - 4 | 1990 | 71 |
AK000667.1 | - | 4 - 5 | 445 | 71 | |
AK295614.1 | - | 4 - 5 | 431 | 71 | |
AK296925.1 | - | 4 - 5 | 410 | 71 | |
AK297468.1 | - | 4 - 5 | 332 | 71 | |
AK308172.1 | - | 4 - 5 | 382 | 71 | |
AY518542.1 | - | 4 - 5 | 344 | 71 | |
AY560593.1 | - | 4 - 5 | 445 | 71 | |
AY560594.1 | - | 4 - 5 | 445 | 71 | |
AY560595.1 | - | 4 - 5 | 445 | 71 | |
AY560596.1 | - | 4 - 5 | 445 | 71 | |
AY560597.1 | - | 4 - 5 | 445 | 71 | |
AY560598.1 | - | 4 - 5 | 445 | 71 | |
AY560599.1 | - | 4 - 5 | 445 | 71 | |
AY560600.1 | - | 4 - 5 | 445 | 71 | |
AY560601.1 | - | 4 - 5 | 445 | 71 | |
AY576417.1 | - | 4 - 5 | 445 | 71 | |
BC014566.2 | - | 4 - 5 | 425 | 71 | |
BT009764.1 | - | 4 - 5 | 344 | 71 | |
U41767.1 | - | 4 - 5 | 351 | 71 | |
U46005.1 | - | 4 - 5 | 344 | 71 |