Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM15
Entrez Gene ID
8751
Gene Name
ADAM metallopeptidase domain 15
Gene Aliases
MDC15
Chromosome Location
Chr.1: 155051272 - 155062776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261464.1 NP_001248393.1 5 - 6 562 81
NM_001261465.1 NP_001248394.1 5 - 6 532 81
NM_001261466.1 NP_001248395.1 5 - 6 484 81
NM_003815.4 NP_003806.3 5 - 6 532 81
NM_207191.2 NP_997074.1 5 - 6 532 81
NM_207194.2 NP_997077.1 5 - 6 532 81
NM_207195.2 NP_997078.1 5 - 6 532 81
NM_207196.2 NP_997079.1 5 - 6 532 81
NM_207197.2 NP_997080.1 5 - 6 532 81
NR_048577.1 5 - 6 532 81
NR_048578.1 5 - 6 532 81
NR_048579.1 5 - 6 532 81
GenBank mRNA AB209157.1 - 4 - 5 2063 81
AK000667.1 - 5 - 6 518 81
AK295614.1 - 5 - 6 504 81
AK296925.1 - 5 - 6 483 81
AK297468.1 - 5 - 6 405 81
AK308172.1 - 5 - 6 455 81
AY518542.1 - 5 - 6 417 81
AY560593.1 - 5 - 6 518 81
AY560594.1 - 5 - 6 518 81
AY560595.1 - 5 - 6 518 81
AY560596.1 - 5 - 6 518 81
AY560597.1 - 5 - 6 518 81
AY560598.1 - 5 - 6 518 81
AY560599.1 - 5 - 6 518 81
AY560600.1 - 5 - 6 518 81
AY560601.1 - 5 - 6 518 81
AY576417.1 - 5 - 6 518 81
BC014566.2 - 5 - 6 498 81
BT009764.1 - 5 - 6 417 81
U41767.1 - 5 - 6 424 81
U46005.1 - 5 - 6 417 81

View Full Product Details