Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF20A
Entrez Gene ID
10112
Gene Name
kinesin family member 20A
Gene Aliases
MKLP2, RAB6KIFL
Chromosome Location
Chr.5: 138178728 - 138187715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005733.2 NP_005724.1 1 - 2 470 71
GenBank mRNA AK025790.1 - 1 - 2 29 71
AK296879.1 - 1 - 2 38 71

View Full Product Details