Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LZTS1
Entrez Gene ID
11178
Gene Name
leucine zipper tumor suppressor 1
Gene Aliases
F37, FEZ1
Chromosome Location
Chr.8: 20246165 - 20303981 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF123655.1 - 3 - 4 1404 78

View Full Product Details