The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005683.3 | NP_005674.2 | 1 - 2 | 59 | 81 |
GenBank mRNA | BC032694.1 | - | 1 - 2 | 59 | 81 |