The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003794.2 | NP_001003794.1 | 6 - 7 | 760 | 90 |
NM_007283.6 | NP_009214.1 | 6 - 7 | 1167 | 90 | |
XM_011512377.1 | XP_011510679.1 | 7 - 8 | 1217 | 90 | |
XM_011512378.1 | XP_011510680.1 | 7 - 8 | 838 | 90 | |
XM_011512379.2 | XP_011510681.1 | 8 - 9 | 954 | 90 | |
XM_011512382.1 | XP_011510684.1 | 6 - 7 | 780 | 90 | |
XM_017005662.1 | XP_016861151.1 | 6 - 7 | 760 | 90 | |
XM_017005663.1 | XP_016861152.1 | 7 - 8 | 831 | 90 | |
XM_017005664.1 | XP_016861153.1 | 7 - 8 | 1328 | 90 | |
XM_017005665.1 | XP_016861154.1 | 5 - 6 | 890 | 90 | |
GenBank mRNA | AJ270950.1 | - | 5 - 6 | 573 | 90 |
AK091314.1 | - | 6 - 7 | 1470 | 90 | |
AK315529.1 | - | 6 - 7 | 807 | 90 | |
AL365376.1 | - | 2 - 3 | 114 | 90 | |
AY172950.1 | - | 4 - 5 | 993 | 90 | |
BC000551.2 | - | 6 - 7 | 752 | 90 | |
BC006230.2 | - | 6 - 7 | 1122 | 90 | |
BX403703.2 | - | 6 - 7 | 679 | 90 | |
BX640777.1 | - | 1 - 2 | 3575 | 90 | |
CR456835.1 | - | 5 - 6 | 603 | 90 | |
U67963.1 | - | 6 - 7 | 753 | 90 |