Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLH3
Entrez Gene ID
27030
Gene Name
mutL homolog 3
Gene Aliases
HNPCC7
Chromosome Location
Chr.14: 75013764 - 75051532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040108.1 NP_001035197.1 12 - 13 4459 69
NM_014381.2 NP_055196.2 11 - 12 4387 69
XM_005267532.4 XP_005267589.1 11 - 12 4354 69
XM_005267533.4 XP_005267590.1 10 - 11 4282 69
XM_006720116.3 XP_006720179.1 12 - 13 4371 69
XM_011536646.2 XP_011534948.1 10 - 11 4288 69
XM_017021219.1 XP_016876708.1 11 - 12 4313 69
GenBank mRNA AB039667.1 - 11 - 12 4243 69
AF195657.1 - 11 - 12 4285 69
AL833875.1 - 11 - 12 2289 69
BC036866.2 - 9 - 10 891 69
BC112167.1 - 10 - 11 4207 69
L40399.1 - 11 - 12 1129 69

View Full Product Details