The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039350.2 | NP_001034439.1 | 12 - 13 | 1695 | 75 |
NM_001290252.1 | NP_001277181.1 | 10 - 11 | 1262 | 75 | |
NM_001936.4 | NP_001927.3 | 12 - 13 | 1397 | 75 | |
NM_130797.3 | NP_570629.2 | 12 - 13 | 1703 | 75 | |
XM_017011808.1 | XP_016867297.1 | 13 - 14 | 1649 | 75 | |
XM_017011809.1 | XP_016867298.1 | 13 - 14 | 1412 | 75 | |
XM_017011810.1 | XP_016867299.1 | 13 - 14 | 1515 | 75 | |
XM_017011811.1 | XP_016867300.1 | 13 - 14 | 1439 | 75 | |
XM_017011812.1 | XP_016867301.1 | 8 - 9 | 810 | 75 | |
GenBank mRNA | AK293582.1 | - | 5 - 6 | 574 | 75 |
AK294483.1 | - | 10 - 11 | 1032 | 75 | |
BC150304.1 | - | 12 - 13 | 1694 | 75 | |
BX647966.1 | - | 12 - 13 | 1460 | 75 | |
M96859.1 | - | 12 - 13 | 1444 | 75 | |
M96860.1 | - | 12 - 13 | 1291 | 75 |