The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039350.2 | NP_001034439.1 | 19 - 20 | 2277 | 87 |
NM_001290252.1 | NP_001277181.1 | 17 - 18 | 1844 | 87 | |
NM_001936.4 | NP_001927.3 | 19 - 20 | 1979 | 87 | |
NM_130797.3 | NP_570629.2 | 19 - 20 | 2285 | 87 | |
XM_017011808.1 | XP_016867297.1 | 20 - 21 | 2231 | 87 | |
XM_017011809.1 | XP_016867298.1 | 20 - 21 | 1994 | 87 | |
XM_017011810.1 | XP_016867299.1 | 20 - 21 | 2097 | 87 | |
XM_017011811.1 | XP_016867300.1 | 20 - 21 | 2021 | 87 | |
XM_017011812.1 | XP_016867301.1 | 15 - 16 | 1392 | 87 | |
GenBank mRNA | AK294483.1 | - | 17 - 18 | 1614 | 87 |
BC150304.1 | - | 19 - 20 | 2276 | 87 | |
BX647966.1 | - | 19 - 20 | 2042 | 87 | |
DA156107.1 | - | 2 - 3 | 175 | 87 | |
M96859.1 | - | 19 - 20 | 2026 | 87 | |
M96860.1 | - | 19 - 20 | 1873 | 87 |