The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039350.2 | NP_001034439.1 | 21 - 22 | 2533 | 83 |
NM_001290252.1 | NP_001277181.1 | 19 - 20 | 2100 | 83 | |
NM_001936.4 | NP_001927.3 | 21 - 22 | 2235 | 83 | |
NM_130797.3 | NP_570629.2 | 21 - 22 | 2541 | 83 | |
XM_017011808.1 | XP_016867297.1 | 22 - 23 | 2487 | 83 | |
XM_017011809.1 | XP_016867298.1 | 22 - 23 | 2250 | 83 | |
XM_017011810.1 | XP_016867299.1 | 22 - 23 | 2353 | 83 | |
XM_017011811.1 | XP_016867300.1 | 22 - 23 | 2277 | 83 | |
XM_017011812.1 | XP_016867301.1 | 17 - 18 | 1648 | 83 | |
GenBank mRNA | AK294483.1 | - | 19 - 20 | 1870 | 83 |
BC150304.1 | - | 21 - 22 | 2532 | 83 | |
BU151391.1 | - | 3 - 4 | 288 | 83 | |
BX647966.1 | - | 21 - 22 | 2298 | 83 | |
DA156107.1 | - | 4 - 5 | 83 | ||
M96859.1 | - | 21 - 22 | 83 | ||
M96860.1 | - | 21 - 22 | 83 |