Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOP3B
Entrez Gene ID
8940
Gene Name
topoisomerase (DNA) III beta
Gene Aliases
TOP3B1
Chromosome Location
Chr.22: 21954380 - 21982843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282112.1 NP_001269041.1 15 - 16 2018 75
NM_001282113.1 NP_001269042.1 15 - 16 2011 75
NM_003935.4 NP_003926.1 15 - 16 2334 75
XM_005261811.1 XP_005261868.1 17 - 18 2184 75
XM_005261813.2 XP_005261870.1 16 - 17 2049 75
XM_006724349.1 XP_006724412.1 16 - 17 1995 75
XM_006724350.1 XP_006724413.1 16 - 17 2169 75
XM_011530482.1 XP_011528784.1 16 - 17 2251 75
XM_017029038.1 XP_016884527.1 15 - 16 1997 75
XM_017029039.1 XP_016884528.1 17 - 18 2059 75
XM_017029040.1 XP_016884529.1 16 - 17 2035 75
XM_017029041.1 XP_016884530.1 17 - 18 2077 75
GenBank mRNA AF017146.1 - 14 - 15 1919 75
AF053082.1 - 14 - 15 1919 75
AF125216.1 - 14 - 15 1879 75
AK096695.1 - 8 - 9 1129 75
AK290997.1 - 15 - 16 1964 75
BC002432.2 - 15 - 16 2241 75
CR456596.1 - 14 - 15 1833 75

View Full Product Details