The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303110.1 | NP_001290039.1 | 3 - 4 | 361 | 106 |
NM_001845.5 | NP_001836.3 | 3 - 4 | 361 | 106 | |
XM_011521048.2 | XP_011519350.1 | 3 - 4 | 8293 | 106 | |
GenBank mRNA | AB209646.1 | - | 3 - 4 | 346 | 106 |
BC142626.1 | - | 3 - 4 | 335 | 106 | |
BC151220.1 | - | 3 - 4 | 345 | 106 | |
CN256264.1 | - | 3 - 4 | 354 | 106 | |
X05561.1 | - | 3 - 4 | 353 | 106 | |
Y00706.1 | - | 1 - 2 | 96 | 106 |