The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303110.1 | NP_001290039.1 | 13 - 14 | 906 | 68 |
NM_001845.5 | NP_001836.3 | 13 - 14 | 906 | 68 | |
XM_011521048.2 | XP_011519350.1 | 13 - 14 | 8838 | 68 | |
GenBank mRNA | AB209646.1 | - | 13 - 14 | 891 | 68 |
BC142626.1 | - | 13 - 14 | 880 | 68 | |
BC151220.1 | - | 13 - 14 | 890 | 68 | |
CD613007.1 | - | 10 - 11 | 553 | 68 | |
X05561.1 | - | 13 - 14 | 898 | 68 | |
Y00706.1 | - | 11 - 12 | 641 | 68 |