The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002887.3 | NP_002878.2 | 6 - 7 | 740 | 82 |
GenBank mRNA | AK222797.1 | - | 6 - 7 | 716 | 82 |
AK302161.1 | - | 5 - 6 | 598 | 82 | |
AK314795.1 | - | 6 - 7 | 728 | 82 | |
BC000528.2 | - | 6 - 7 | 718 | 82 | |
BC014619.2 | - | 6 - 7 | 718 | 82 | |
BT007394.1 | - | 6 - 7 | 699 | 82 | |
S80343.1 | - | 6 - 7 | 726 | 82 |